missing translation for 'onlineSavingsMsg'
Learn More

Guanosine 5'-Monophosphate Disodium Salt, TRC

Product Code. 17998674
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17998674 5 g 5g
17997634 1 g 1g
17969654 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17998674 Supplier Toronto Research Chemicals Supplier No. TRCS6341005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146894-16-8
Molecular Formula C10H12N5Na2O8P • x(H2O)
Molecular Weight (g/mol) 407.181802
Synonym 5'-Guanylic Acid Disodium Salt, 5'-GMP Disodium Salt, Disodium 5'-GMP, Disodium 5'-Guanylate, Disodium GMP, Disodium Guanosine 5'-Monophosphate, Disodium Guanosine 5'-phosphate, Disodium Guanylate, E 627, GMP Disodium Salt, GMP Sodium Salt, Guanosine 5'-Monophosphate Disodium Salt, Guanosine 5'-Phosphate Disodium Salt, Guanylic Acid Sodium Salt, NSC 20265, Sodium 5'-Guanylate, Sodium GMP, Sodium Guanosine 5'-Monophosphate;
SMILES O[C@H]1[C@@H](O)[C@H](N2C(NC(N)=NC3=O)=C3N=C2)O[C@@H]1COP(O[Na])(O[Na])=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H14N5O8P.2Na.H2O/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(23-9)1-22-24(19,20)21;;;/h2-3,5-6,9,16-17H,1H2,(H2,19,20,21)(H3,11,13,14,18);;;1H2/q;2*+1;/p-2/t3-,5-,6-,9-;;;/m1.../s1
Formula Weight 425.0325
Chemical Name or Material Guanosine 5'-Monophosphate Disodium Salt Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.