CAS RN 146894-16-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Guanosine 5'-Monophosphate Disodium Salt, TRC
CAS: 146894-16-8 Molecular Formula: C10H12N5Na2O8P • x(H2O) Molecular Weight (g/mol): 407.181802 Synonym: 5'-Guanylic Acid Disodium Salt,5'-GMP Disodium Salt,Disodium 5'-GMP,Disodium 5'-Guanylate,Disodium GMP,Disodium Guanosine 5'-Monophosphate,Disodium Guanosine 5'-phosphate,Disodium Guanylate,E 627,GMP Disodium Salt,GMP Sodium Salt,Guanosine 5'-Monophosphate Disodium Salt,Guanosine 5'-Phosphate Disodium Salt,Guanylic Acid Sodium Salt,NSC 20265,Sodium 5'-Guanylate,Sodium GMP,Sodium Guanosine 5'-Monophosphate; SMILES: O[C@H]1[C@@H](O)[C@H](N2C(NC(N)=NC3=O)=C3N=C2)O[C@@H]1COP(O[Na])(O[Na])=O