Learn More
Guanidinoethyl Sulfonate, TRC
Product Code. 30240814
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240814 1 g 1g
30240774 100 mg 100mg
30240574 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30240814 Supplier Toronto Research Chemicals Supplier No. TRCG8275001G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 543-18-0
Molecular Formula C3 H9 N3 O3 S
Molecular Weight (g/mol) 167.19
IUPAC Name 2-carbamimidamidoethanesulfonic acid
SMILES NC(=N)NCCS(=O)(=O)O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C3H9N3O3S/c4-3(5)6-1-2-10(7,8)9/h1-2H2,(H4,4,5,6)(H,7,8,9)
Formula Weight 167.0365
Percent Purity >95%
Chemical Name or Material Guanidinoethyl Sulfonate
Grade HPLC