accessibility menu, dialog, popup

UserName

CAS RN 543-18-0

NH2NH2NSHOOO

CAS RN 543-18-0

IUPAC Name: 2-[(diaminomethylidene)amino]ethane-1-sulfonic acid
Synonyms: 2-[(diaminomethylidene)amino]ethanesulfonic acid
Molecular Weight (g/mol): 167.18
Molecular Formula: C3H9N3O3S
InChi Key: JKLRIMRKZBSSED-UHFFFAOYSA-N
SMILES: NC(N)=NCCS(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results