missing translation for 'onlineSavingsMsg'
Learn More

Guanidinoethyl Sulfonate, TRC

Product Code. 30240774
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240774 100 mg 100mg
30240814 1 g 1g
30240574 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240774 Supplier Toronto Research Chemicals Supplier No. TRCG827500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-18-0
Molecular Formula C3 H9 N3 O3 S
Molecular Weight (g/mol) 167.19
IUPAC Name 2-carbamimidamidoethanesulfonic acid
SMILES NC(=N)NCCS(=O)(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C3H9N3O3S/c4-3(5)6-1-2-10(7,8)9/h1-2H2,(H4,4,5,6)(H,7,8,9)
Formula Weight 167.0365
Percent Purity >95%
Chemical Name or Material Guanidinoethyl Sulfonate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.