missing translation for 'onlineSavingsMsg'
Learn More

GRWD1 Antibody, Novus Biologicals™

Product Code. 18214800 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214800 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214800 Supplier Novus Biologicals Supplier No. NB10077320
  • ONLINE EXCLUSIVE PRICE
    583.00 EUR
    551.25 EUR / 0.1mL
    Save 31.75 EUR
    5% Off
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GRWD1 Polyclonal specifically detects GRWD1 in Human samples. It is validated for Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRWD1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_113673.2
Gene Alias CDW4, CUL4- and DDB1-associated WDR protein 4, glutamate-rich WD repeat containing 1, GRWDglutamate rich WD repeat protein GRWD, KIAA1942regulator of ribosome biogenesis 1 homolog, RRB1, WD repeat domain 28, WDR28glutamate-rich WD repeat-containing protein 1
Gene Symbols GRWD1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 350 and 400 of human glutamate-rich WD repeat containing 1 (WD repeat domain 28) using the numbering given in entry NP_113673.2 (GeneID 83743).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83743
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.