missing translation for 'onlineSavingsMsg'
Learn More

GRP78/HSPA5 Antibody (3C5-1A4), FITC, Novus Biologicals™

Product Code. 18214774 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214774 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214774 Supplier Novus Biologicals Supplier No. NBP259692F
  • ONLINE EXCLUSIVE PRICE
    513.00 EUR
    485.10 EUR / 100µL
    Save 27.90 EUR
    5% Off
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GRP78/HSPA5 Monoclonal specifically detects GRP78/HSPA5 in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRP78/HSPA5
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3C5-1A4
Conjugate FITC
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias 78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2
Gene Symbols HSPA5
Host Species Mouse
Immunogen Full-length recombinant rat GRP78
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR)
Primary or Secondary Primary
Gene ID (Entrez) 3309
Test Specificity Detects approx 33 kDa protein.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.