missing translation for 'onlineSavingsMsg'
Learn More

Gr-1/Ly-6G Antibody (1A8), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18215040 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18215040 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18215040 Supplier Novus Biologicals Supplier No. NBP253131AF488
  • ONLINE EXCLUSIVE PRICE
    355.00 EUR
    334.95 EUR / 0.1mL
    Save 20.05 EUR
    5% Off
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rat Monoclonal Antibody

Gr-1/Ly-6G Monoclonal specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gr-1/Ly-6G
Applications Flow Cytometry
Classification Monoclonal
Clone 1A8
Conjugate Alexa Fluor 488
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C6orf23, G6D, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25
Gene Symbols Ly6g
Host Species Rat
Immunogen Ly-6G transfected EL-4J cell line.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58530
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.