missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Rabbit IgG (H+L) Secondary Antibody, FITC (Pre-adsorbed), Novus Biologicals™

Product Code. 18214793 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214793 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214793 Supplier Novus Biologicals Supplier No. NBP173136
  • ONLINE EXCLUSIVE PRICE
    311.00 EUR
    294.00 EUR / 2mg
    Save 17.00 EUR
    5% Off
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Goat Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Fluorescein
Dilution Western Blot 1:1000 - 1:10000, Flow Cytometry, Immunocytochemistry/Immunofluorescence 1:500 - 1:2500
Host Species Goat
Immunogen Rabbit IgG whole molecule
Purification Method Affinity Purified
Quantity 2 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity This antibody was pre-adsorbed against Human Serum Proteins. No reaction was observed against Human Serum Proteins.
Reconstitution Reconstitute with 1.0 ml deionized water (or equivalent).
Target Species Rabbit
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.