missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Guinea Pig IgG (H+L) Secondary Antibody, FITC, Novus Biologicals™

Product Code. 18215122 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18215122 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18215122 Supplier Novus Biologicals Supplier No. NB7397
  • ONLINE EXCLUSIVE PRICE
    261.00 EUR
    246.75 EUR / 1mL
    Save 14.25 EUR
    5% Off
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Goat Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Guinea Pig samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate FITC
Dilution Flow Cytometry 1:50 - 1:200, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunohistochemistry-Paraffin
Host Species Goat
Immunogen Guinea Pig IgG-heavy and light chain
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity By immunoelectrophoresis and ELISA this antibody reacts specifically with guinea pig IgG and with light chains common to other guinea pig immunoglobulins. No antibody was detected against nonimmunoglobulin serum proteins. This antibody may cross react with IgG from other species.
Target Species Guinea Pig
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.