missing translation for 'onlineSavingsMsg'
Learn More

Glycoaldehyde Dimer, TRC

Product Code. 30133548
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133548 25 g 25g
30132540 5 g 5g
17962635 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133548 Supplier Toronto Research Chemicals Supplier No. TRCG64115025G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23147-58-2
Molecular Formula C4H8O4
Molecular Weight (g/mol) 120.1
Synonym p-Dioxane-2,5-diol, 2,5-Dihydroxy-1,4-dioxane, 2,5-Dihydroxy-p-dioxane
IUPAC Name 1,4-dioxane-2,5-diol
SMILES OC1OCC(O)OC1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C4H8O4/c5-3-1-7-4(6)2-8-3/h3-6H,1-2H2
Formula Weight 120.0423
Chemical Name or Material Glycoaldehyde Dimer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.