CAS RN 23147-58-2
CAS RN 23147-58-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Glycoaldehyde Dimer, TRC
CAS: 23147-58-2 Molecular Formula: C4H8O4 Molecular Weight (g/mol): 120.1 Synonym: p-Dioxane-2,5-diol,2,5-Dihydroxy-1,4-dioxane,2,5-Dihydroxy-p-dioxane IUPAC Name: 1,4-dioxane-2,5-diol SMILES: OC1OCC(O)OC1