missing translation for 'onlineSavingsMsg'
Learn More

GLIPR2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18214919 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214919 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214919 Supplier Novus Biologicals Supplier No. NBP2849760.1ML
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GLIPR2 Polyclonal specifically detects GLIPR2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLIPR2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias C9orf19Glioma pathogenesis-related protein 2, chromosome 9 open reading frame 19, GAPR1, GAPR-117kD fetal brain protein, GLI pathogenesis-related 2, GliPR 2, Golgi-associated plant pathogenesis-related protein 1, Golgi-associated PR-1 protein
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human GLIPR2
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152007
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.