Learn More
MedChemExpress Ginkgetin
Product Code. 16441578
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16441578 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16441578 Supplier MedChemExpress Supplier No. HYN0889
 Restricted Item
Estimated Shipment: 16-11-2026
MedChemExpress Ginkgetin, a biflavone, is isolated from Ginkgo biloba leaves. Ginkgetin exhibit anti-tumor, anti-inflammatory, neuroprotective, anti-fungal activities. Ginkgetin is also a potent inhibitor of Wnt signaling, with an IC50 of 5.92 μμ.
Specifications
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 566.51 g/mol
Shipping Condition Room temperature
For Use With (Application) COVID-19-immunoregulation
CAS 481-46-9
Product Type Natural Products
Purity 97.66%
Solubility DMSO : 125 mg/mL (220.65 mM∣Need ultrasonic)
Target Wnt∣Apoptosis∣Autophagy
Quantity 5 mg