Learn More
Gilteritinib, TRC
Product Code. 30137852
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137852 100 mg 100mg
30132424 50 mg 50mg
30010404 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137852 Supplier Toronto Research Chemicals Supplier No. TRCG377700100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1254053-43-4
Molecular Formula C29H44N8O3
Molecular Weight (g/mol) 552.71
Synonym ASP 2215, 6-Ethyl-3-[[3-methoxy-4-[4-(4-methyl-1-piperazinyl)-1-piperidinyl]phenyl]amino]-5-[(tetrahydro-2H-pyran-4-yl)amino]-2-pyrazinecarboxamide
IUPAC Name 6-ethyl-3-[3-methoxy-4-[4-(4-methylpiperazin-1-yl)piperidin-1-yl]anilino]-5-(oxan-4-ylamino)pyrazine-2-carboxamide
SMILES NC(C(N=C1CC)=C(NC(C=C2)=CC(OC)=C2N3CCC(CC3)N4CCN(CC4)C)N=C1NC5CCOCC5)=O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C29H44N8O3/c1-4-23-28(31-20-9-17-40-18-10-20)34-29(26(33-23)27(30)38)32-21-5-6-24(25(19-21)39-3)37-11-7-22(8-12-37)36-15-13-35(2)14-16-36/h5-6,19-20,22H,4,7-18H2,1-3H3,(H2,30,38)(H2,31,32,34)
Formula Weight 552.3536
Chemical Name or Material Gilteritinib