missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Geminin Monoclonal specifically detects Geminin in Human samples. It is validated for ELISA, Western Blot.
Specifications
Specifications
| Antigen | Geminin |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2H7 |
| Conjugate | Alexa Fluor 350 |
| Dilution | Western Blot, ELISA |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | Gem, geminin, geminin, DNA replication inhibitor, RP3-369A17.3 |
| Gene Symbols | GMNN |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?