Learn More
Cytiva Gel Filtration Calibration Kits
Product Code. 10734557 Shop All Cytiva Products
Change view
Click to view available options
MWCO:
43,000 to 669,000
6500 to 75,000
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Unit Size
10734557 6500 to 75,000 1 set
10196234 43,000 to 669,000 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10734557 Supplier Cytiva Supplier No. 28403841
 Restricted Item
In Stock
Calibrate gel filtration columns simply, reliably. Cytiva Life Sciences™ Gel Filtration Calibration Kits contain a well-defined series of protein standards.
  • Optimized range of proteins suited to high-resolution media and prepacked columns
  • Each kit contains five vials of lyophilized proteins
  • Blue Dextran 2000 included to determine the void fraction of the column
TRUSTED_SUSTAINABILITY
Specifications
Quantity Each
For Use With (Equipment) High-resolution media and prepacked column
Product Type Calibration Kit
For Use With (Application) Calibration of high-resolution gel filtration media and column such as Superdex™, Superose and Sephacryl
Includes 10mg Aprotinin (6500 Mr), 50mg Ribonuclease A (13,700 Mr), 15mg Carbonic anhydrase (29,000 Mr), 50mg Ovalbumin (43,000 Mr), 50mg Conalbumin (75,000 Mr), 50mg Blue Dextran (2,000,000 Mr)
MWCO 6500 to 75,000
Content And Storage 2° to 8°C
compliance-icons
Product Identifier
  • Gel Filtration Calibration Kit LMW
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Trypsin inhibitor, pancreatic basic