missing translation for 'onlineSavingsMsg'
Learn More

Fluoxastrobin, TRC

Product Code. 30138453
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
100mg
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138453 250 mg 250mg
30134880 2.5 g 2.5g
17934835 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138453 Supplier Toronto Research Chemicals Supplier No. TRCF596700250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 361377-29-9
Molecular Formula C21 H16 Cl F N4 O5
Molecular Weight (g/mol) 458.83
Synonym Disarm, Disarm 480SC, Evito, Evito 480SC, Fluoxastrobin, Fluoxastrobin ST, HEC 5725, (1E)-[2-[[6-(2-Chlorophenoxy)-5-fluoro-4-pyrimidinyl]oxy]phenyl](5,6-dihydro-1,4,2-dioxazin-3-yl)methanone O-methyloxime
IUPAC Name (E)-1-[2-[6-(2-chlorophenoxy)-5-fluoropyrimidin-4-yl]oxyphenyl]-1-(5,6-dihydro-1,4,2-dioxazin-3-yl)-N-methoxymethanimine
SMILES CO\N=C(\C1=NOCCO1)/c2ccccc2Oc3ncnc(Oc4ccccc4Cl)c3F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C21H16ClFN4O5/c1-28-26-18(21-27-30-11-10-29-21)13-6-2-4-8-15(13)31-19-17(23)20(25-12-24-19)32-16-9-5-3-7-14(16)22/h2-9,12H,10-11H2,1H3/b26-18+
Formula Weight 458.0793
Chemical Name or Material Fluoxastrobin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.