CAS RN 361377-29-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fluoxastrobin, TRC
CAS: 361377-29-9 Molecular Formula: C21 H16 Cl F N4 O5 Molecular Weight (g/mol): 458.83 Synonym: Disarm,Disarm 480SC,Evito,Evito 480SC,Fluoxastrobin,Fluoxastrobin ST,HEC 5725,(1E)-[2-[[6-(2-Chlorophenoxy)-5-fluoro-4-pyrimidinyl]oxy]phenyl](5,6-dihydro-1,4,2-dioxazin-3-yl)methanone O-methyloxime IUPAC Name: (E)-1-[2-[6-(2-chlorophenoxy)-5-fluoropyrimidin-4-yl]oxyphenyl]-1-(5,6-dihydro-1,4,2-dioxazin-3-yl)-N-methoxymethanimine SMILES: CO\N=C(\C1=NOCCO1)/c2ccccc2Oc3ncnc(Oc4ccccc4Cl)c3F