Learn More
Tocris Bioscience™ FHT 2344
Description
FHT 2344 is a potent allosteric and ATP-competitive inhibitor of SMARCA2/SMARCA4 ATPases (also known as BRM/BRG1) of the BAF complex (IC50 values are 13.8 and 26.1 nM at SMARCA2 and SMARCA4, respectively). FHT 2344 inhibits SMARCA2 transcriptional activity in a BRG1-mutant cell line and SMARCA4 transcription in a BRM-mutant cell line (IC50 values are 29.8 and 30.2 nM, respectively). Dose-dependently reduces tumor growth in a uveal melanoma xenograft model and suppresses SOX10 transcription.
Specifications
Specifications
| Chemical Name or Material | N-[(1S)-1-(Methoxymethyl)-2-[[4-[3-(1-methyl-1H-pyrazol-3-yl)phenyl]-2-thiazolyl]amino]-2-oxoethyl]-1-(methylsulfonyl)-1H-pyrrole-3-carboxamide |
| CAS | 2468058-90-2 |
| Quantity | 10 mg |
| Target | Bromodomain Inhibitors |
| Molecular Formula | C23H24N6O5S2 |
| Purity | 0.95 |