missing translation for 'onlineSavingsMsg'
Learn More

Fenpropimorph, TRC

Product Code. 30137100
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137100 250 mg 250mg
17961505 10 mg 10mg
17912625 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137100 Supplier Toronto Research Chemicals Supplier No. TRCF249500250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67564-91-4
Molecular Formula C20 H33 N O
Molecular Weight (g/mol) 303.48
Synonym Morpholine, 4-[3-[4-(1,1-dimethylethyl)phenyl]-2-methylpropyl]-2,6-dimethyl-, cis-, rel-(2R,6S)-4-[3-[4-(1,1-Dimethylethyl)phenyl]-2-methylpropyl]-2,6-dimethylmorpholine, BAS 42100F, Corbel, Fenpropimorph, Forbel 750, Mistral, Volley 88OL, cis-4-[3-(4-tert-Butylphenyl)-2-methylpropyl]-2,6-dimethylmorpholine
IUPAC Name (2R,6S)-4-[3-(4-tert-butylphenyl)-2-methylpropyl]-2,6-dimethylmorpholine
SMILES CC(CN1C[C@@H](C)O[C@@H](C)C1)Cc2ccc(cc2)C(C)(C)C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C20H33NO/c1-15(12-21-13-16(2)22-17(3)14-21)11-18-7-9-19(10-8-18)20(4,5)6/h7-10,15-17H,11-14H2,1-6H3/t15?,16-,17+
Formula Weight 303.2562
Percent Purity >95%
Chemical Name or Material Fenpropimorph
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.