accessibility menu, dialog, popup

UserName

CAS RN 67564-91-4

CH3N(S)CH3O(R)H3CCH3CH3H3C

CAS RN 67564-91-4

IUPAC Name: (2R,6S)-4-[3-(4-tert-butylphenyl)-2-methylpropyl]-2,6-dimethylmorpholine
Synonyms: fenpropimorph fenpropimorph (iso) fenpropimorph cis-form cis-fenpropimorph fenpropimorphe
Molecular Weight (g/mol): 303.49
Molecular Formula: C20H33NO
InChi Key: RYAUSSKQMZRMAI-ALOPSCKCSA-N
SMILES: CC(CN1C[C@H](C)O[C@H](C)C1)CC1=CC=C(C=C1)C(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (4)
Concentration 
  • (2)
  • (2)
Solvent or Matrix 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results