Learn More
Fenpropidin, TRC
Product Code. 30393098
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393098 100 mg
30398295 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30393098 Supplier Toronto Research Chemicals Supplier No. TRCF249470100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 67306-00-7
Molecular Formula C19 H31 N
Molecular Weight (g/mol) 273.46
Synonym 1-[3-[4-(1,1-Dimethylethyl)phenyl]-2-methylpropyl]piperidine, F 3240, Fenpropidin, Fenpropidine, Mildin, Patrol, Piperidine, 1-[3-[4-(1,1-dimethylethyl)phenyl]-2-methylpropyl]-
IUPAC Name 1-[3-(4-tert-butylphenyl)-2-methylpropyl]piperidine
SMILES CC(CN1CCCCC1)Cc2ccc(cc2)C(C)(C)C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C19H31N/c1-16(15-20-12-6-5-7-13-20)14-17-8-10-18(11-9-17)19(2,3)4/h8-11,16H,5-7,12-15H2,1-4H3
Formula Weight 273.2457
Percent Purity >95%
Chemical Name or Material Fenpropidin
Grade GC