accessibility menu, dialog, popup

UserName

CAS RN 67306-00-7

CH3NCH3CH3H3C

CAS RN 67306-00-7

IUPAC Name: 1-[3-(4-tert-butylphenyl)-2-methylpropyl]piperidine
Synonyms: fenpropidin
Molecular Weight (g/mol): 273.46
Molecular Formula: C19H31N
InChi Key: MGNFYQILYYYUBS-UHFFFAOYNA-N
SMILES: CC(CN1CCCCC1)CC1=CC=C(C=C1)C(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
Concentration 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results