missing translation for 'onlineSavingsMsg'
Learn More

Fenoxanil, TRC

Product Code. 30138607
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138607 250 mg 250mg
17954855 50 mg 50mg
17954935 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138607 Supplier Toronto Research Chemicals Supplier No. TRCF248888250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115852-48-7
Molecular Formula C15 H18 Cl2 N2 O2
Molecular Weight (g/mol) 329.22
Synonym N-(1-Cyano-1,2-dimethylpropyl)-2-(2,4-dichlorophenoxy)propanamide, AC 382042, AC 901216, CL 382042, Fenoxanil, Magnaporthe amide, NNF 9425, WL 378309
IUPAC Name N-(2-cyano-3-methylbutan-2-yl)-2-(2,4-dichlorophenoxy)propanamide
SMILES CC(C)C(C)(NC(=O)C(C)Oc1ccc(Cl)cc1Cl)C#N

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H18Cl2N2O2/c1-9(2)15(4,8-18)19-14(20)10(3)21-13-6-5-11(16)7-12(13)17/h5-7,9-10H,1-4H3,(H,19,20)
Formula Weight 328.0745
Chemical Name or Material Fenoxanil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.