CAS RN 115852-48-7
CAS RN 115852-48-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fenoxanil, TRC
CAS: 115852-48-7 Molecular Formula: C15 H18 Cl2 N2 O2 Molecular Weight (g/mol): 329.22 Synonym: N-(1-Cyano-1,2-dimethylpropyl)-2-(2,4-dichlorophenoxy)propanamide,AC 382042,AC 901216,CL 382042,Fenoxanil,Magnaporthe amide,NNF 9425,WL 378309 IUPAC Name: N-(2-cyano-3-methylbutan-2-yl)-2-(2,4-dichlorophenoxy)propanamide SMILES: CC(C)C(C)(NC(=O)C(C)Oc1ccc(Cl)cc1Cl)C#N