Learn More
Fasentin, TRC
Product Code. 30402652
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402652 25 mg
30396934 10 mg
30390594 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30402652 Supplier Toronto Research Chemicals Supplier No. TRCF15002025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 392721-37-8
Molecular Formula C11H9ClF3NO2
Molecular Weight (g/mol) 279.64
Synonym N-(4-Chloro-3-(trifluoromethyl)phenyl)-3-oxobutanamide
IUPAC Name N-[4-chloro-3-(trifluoromethyl)phenyl]-3-oxobutanamide
SMILES CC(CC(NC1=CC(C(F)(F)F)=C(Cl)C=C1)=O)=O
Spezifikation
Quantity 25 mg
InChI Formula InChI=1S/C11H9ClF3NO2/c1-6(17)4-10(18)16-7-2-3-9(12)8(5-7)11(13,14)15/h2-3,5H,4H2,1H3,(H,16,18)
Formula Weight 279.0274
Chemical Name or Material Fasentin