accessibility menu, dialog, popup

UserName

CAS RN 392721-37-8

H3COOHNClFFF

CAS RN 392721-37-8

IUPAC Name: N-[4-chloro-3-(trifluoromethyl)phenyl]-3-oxobutanamide
Synonyms: N-[4-chloro-3-(trifluoromethyl)phenyl]-3-oxobutanamide
Molecular Weight (g/mol): 279.64
Molecular Formula: C11H9ClF3NO2
InChi Key: GNYIJZMBLZXJEJ-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)NC1=CC(=C(Cl)C=C1)C(F)(F)F
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results