missing translation for 'onlineSavingsMsg'
Learn More

Famoxadone, TRC

Product Code. 30135578
Change view
Click to view available options
Quantity:
250 mg
1 g
50 g
Unit Size:
1g
250mg
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135578 50 g 50g
17941835 1 g 1g
17914855 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135578 Supplier Toronto Research Chemicals Supplier No. TRCF10230050G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131807-57-3
Molecular Formula C22 H18 N2 O4
Molecular Weight (g/mol) 374.39
Synonym 5-Methyl-5-(4-phenoxyphenyl)-3-(phenylamino)-2,4-oxazolidinedione, DPX-JE 874, Famoxadone, Famoxate, 2,4-Oxazolidinedione, 5-methyl-5-(4-phenoxyphenyl)-3-(phenylamino)-
IUPAC Name 3-anilino-5-methyl-5-(4-phenoxyphenyl)-1,3-oxazolidine-2,4-dione
SMILES CC1(OC(=O)N(Nc2ccccc2)C1=O)c3ccc(Oc4ccccc4)cc3

Specifications

Quantity 50 g
InChI Formula InChI=1S/C22H18N2O4/c1-22(16-12-14-19(15-13-16)27-18-10-6-3-7-11-18)20(25)24(21(26)28-22)23-17-8-4-2-5-9-17/h2-15,23H,1H3
Formula Weight 374.1267
Percent Purity >95%
Chemical Name or Material Famoxadone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.