accessibility menu, dialog, popup

UserName

CAS RN 131807-57-3

H3COONHNOO

CAS RN 131807-57-3

IUPAC Name: 5-methyl-5-(4-phenoxyphenyl)-3-(phenylamino)-1,3-oxazolidine-2,4-dione
Synonyms: famoxadone
Molecular Weight (g/mol): 374.4
Molecular Formula: C22H18N2O4
InChi Key: PCCSBWNGDMYFCW-UHFFFAOYNA-N
SMILES: CC1(OC(=O)N(NC2=CC=CC=C2)C1=O)C1=CC=C(OC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results