missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FAM105A Polyclonal specifically detects FAM105A in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | FAM105A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | Q9NUU6 |
| Gene Alias | family with sequence similarity 105, member A, FLJ11127, FLJ43660, hypothetical protein LOC54491, NET20 |
| Gene Symbols | FAM105A |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?