Learn More
Thermo Scientific™ EZ-Link™ Sulfo-NHS-Biotin and Labeling Kits
Product Code. 10773495 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
8 x 1 mg
Unit Size:
8mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10773495 8 x 1 mg 8mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10773495 Supplier Thermo Scientific™ Supplier No. 21326
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Biotinylate protein amines (e.g., lysines) with our shortest (13.5A), water-soluble, NHS-ester biotinylation reagent for antibody or cell-surface labeling.
Label antibodies, proteins and other molecules that have primary amines with Thermo Scientific™ EZ-Link Sulfo-NHS-Biotin, a short-chain, water-soluble biotinylation reagent.
Highlights:
  • Shorter spacer arm than Sulfo-NHS-LC-Biotin
  • Reacts with primary amines at pH 7-9 in non-amine-containing buffers such as borate, carbonate, or phosphate buffers
  • Negative charge of -SO3 keeps biotinylation localized at the cell surface so reagent does not pass through cell membrane
  • Water solubility eliminates the need for harmful organic solvents
U.S. Patents 5,892,628 and 5,872,261.
TRUSTED_SUSTAINABILITY
Specifications
Cleavable No
Description EZ-Link Sulfo-NHS-Biotin, No-Weigh Format
Formulation Sulfo-NHS ester of biotin
Molecular Weight (g/mol) 443.43
Membrane Permeable No
PEGylated No
Spacer Arm Length 13.5
Quantity 8 x 1 mg
Sufficient For 8 uses to label a total of 120mg of IgG using typical conditions
Content And Storage -20°C with desiccant
Chemical Reactivity Amine
Solubility Water
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.