Learn More
Thermo Scientific™ EZ-Link™ NHS-PEG12-Biotin, No-Weigh™ Format
Product Code. 11801195 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mg
10 x 1 mg
500 mg
Unit Size:
10mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11801195 25 mg 25mg
15792029 10 x 1 mg 10mg
11811195 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11801195 Supplier Thermo Scientific™ Supplier No. 21312
  • ONLINE EXCLUSIVE PRICE
    363.00 EUR
    272.65 EUR / 25mg
    Save 90.35 EUR
    24% Off
 Restricted Item
In Stock
Biotinylate protein amines and maximize solubility of proteins with this very long (56.0A) NHS-ester biotinylation reagent containing a pegylated, water-soluble spacer arm.
Thermo Scientific™ EZ-Link NHS-PEG12-Biotin is very long, pegylated biotinylation reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.
Highlights:
  • Water-soluble polyethylene glycol (PEG) spacer arm increases solubility of labeled protein
  • Excellent purity, molecular weight uniformity and amine-reactivity
  • Long spacer arm reduces steric hindrance when binding to avidin molecules
TRUSTED_SUSTAINABILITY
Specifications
Product Type NHS-PEG12-Biotin
Quantity 25 mg
Content And Storage Store desiccated at -20°C. Shipped at ambient temperature.
Product Line EZ-Link
Cell Permeability Cell-Impermeant
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Biotin
Spacer Extra-long, Pegylated
Label Type Biotin & Analogs