Learn More
Thermo Scientific™ Exonuclease III (200 U/μL)
Product Code. 10858990 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
4,000 units
Unit Size:
4000 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10858990 4,000 units 4000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10858990 Supplier Thermo Scientific™ Supplier No. EN0191
  • ONLINE EXCLUSIVE PRICE
    39.57 EUR
    37.65 EUR / Pack of 4000
    Save 1.92 EUR
    4% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Digest double-stranded DNA or DNA-RNA hybrids with Exonuclease III is a 3'–>5' exonuclease.
Exonuclease III (ExoIII) exhibits four catalytic activities (see Reference1). The 3'≥5' exodeoxyribonuclease activity of ExoIII is specific for double-stranded DNA. ExoIII degrades dsDNA from blunt ends, 5'-overhangs or nicks, releases 5'-mononucleotides from the 3'-ends of DNA strands and produces stretches of single-stranded DNA. It is not active on 3'-overhang ends of DNA that are at least four-bases long and do not carry a 3'-terminal C-residue (see Reference 2) on single-stranded DNA, or on phosphorothioate-linked nucleotides.
ExoIII 3'-phosphatase activity removes the 3'-terminal phosphate, generating a 3'-OH group. ExoIII Rnase H activity exonucleolytically degrades the RNA strand in RNA-DNA hybrids. ExpOOO apurinic/apyrimidinic-endonuclease activity cleaves phosphodiester bonds at apurinic or apyrimidinic sites to produce 5'-termini that are base-free deoxyribose 5'-phosphate residues.
Highlights
  • Active in restriction enzyme buffers
TRUSTED_SUSTAINABILITY
Specifications
Concentration 200 U/μL
Enzyme Exonuclease
Compatible Buffer Enzyme Buffer
Quantity 4,000 units
Product Type Exonuclease III