missing translation for 'onlineSavingsMsg'
Learn More

Etofenprox, TRC

Product Code. 30135834
Change view
Click to view available options
Quantity:
100 mg
5 g
10 g
Unit Size:
100mg
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135834 5 g 5g
30132959 10 g 10g
17915005 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135834 Supplier Toronto Research Chemicals Supplier No. TRCE9364005G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80844-07-1
Molecular Formula C25 H28 O3
Molecular Weight (g/mol) 376.49
Synonym Benzene, 1-[[2-(4-ethoxyphenyl)-2-methylpropoxy]methyl]-3-phenoxy-, 1-[[2-(4-Ethoxyphenyl)-2-methylpropoxy]methyl]-3-phenoxybenzene, 2-(4-Ethoxyphenyl)-2-methylpropyl 3-phenoxybenzyl ether, 4-Ethoxyneophyl 3-phenoxybenzyl ether, Ethofenprox, Ethophenprox, Ethoproxyfen, Ethoproxyphen, Etof, Etofenprox, MTI 500, Myungtaja, SA 130301, Safety (insecticide), Trebon, Trebon Star, Vectron, Vectron 20WP, Zenivex E 20
IUPAC Name 1-ethoxy-4-[2-methyl-1-[(3-phenoxyphenyl)methoxy]propan-2-yl]benzene
SMILES CCOc1ccc(cc1)C(C)(C)COCc2cccc(Oc3ccccc3)c2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C25H28O3/c1-4-27-22-15-13-21(14-16-22)25(2,3)19-26-18-20-9-8-12-24(17-20)28-23-10-6-5-7-11-23/h5-17H,4,18-19H2,1-3H3
Formula Weight 376.2038
Percent Purity >95%
Chemical Name or Material Etofenprox
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.