accessibility menu, dialog, popup

UserName

CAS RN 80844-07-1

CH3OH3CCH3OO

CAS RN 80844-07-1

IUPAC Name: 1-ethoxy-4-{2-methyl-1-[(3-phenoxyphenyl)methoxy]propan-2-yl}benzene
Synonyms: trebon ethofenprox ethophenprox etofenproxum etofenprox
Molecular Weight (g/mol): 376.5
Molecular Formula: C25H28O3
InChi Key: YREQHYQNNWYQCJ-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)C(C)(C)COCC1=CC=CC(OC2=CC=CC=C2)=C1
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results