Learn More
Ethyl (trimethylsilyl)acetate, 98%
Product Code. 11453288
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453288 10 g 10g
11463288 50 g 50g
11473288 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11453288 Supplier Thermo Scientific Alfa Aesar Supplier No. A17707.09
 Restricted Item
In Stock
Add to basket
CAS: 4071-88-9 | C7H16O2Si | 160.288 g/mol
Ethyl trimethylsilylacetate is used in the synthesis of ethyl-2,2-dibromo-2-(trimethylsilyl)acetate. It was also used to silylate the enolizable aldehydes and ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl trimethylsilylacetate is used in the synthesis of ethyl-2,2-dibromo-2-(trimethylsilyl)acetate. It was also used to silylate the enolizable aldehydes and ketones.
Solubility
Decomposes in water.
Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4071-88-9
Molecular Formula C7H16O2Si
Molecular Weight (g/mol) 160.288
MDL Number MFCD00009172
InChI Key QQFBQBDINHJDMN-UHFFFAOYSA-N
Synonym ethyl trimethylsilyl acetate, ethyl trimethylsilylacetate, ethyl 2-trimethylsilyl acetate, etsa, trimethylsilyl acetic acid ethyl ester, acetic acid, trimethylsilyl-, ethyl ester, pubchem16058, acmc-1ahie
PubChem CID 77687
IUPAC Name ethyl 2-trimethylsilylacetate
SMILES CCOC(=O)C[Si](C)(C)C
Specifications
Density 0.876
Boiling Point 156°C to 158°C
Flash Point 35°C (95°F)
Refractive Index 1.415
Quantity 10 g
UN Number UN3272
Beilstein 1755902
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 160.29
Percent Purity ≥98%
Chemical Name or Material Ethyl (trimethylsilyl)acetate