accessibility menu, dialog, popup

UserName

CH3OOSiCH3CH3H3C

CAS RN 4071-88-9

IUPAC Name: ethyl 2-(trimethylsilyl)acetate
Synonyms: ethyl 2-(trimethylsilyl)acetate
Molecular Weight (g/mol): 160.29
Molecular Formula: C7H16O2Si
InChi Key: QQFBQBDINHJDMN-UHFFFAOYSA-N
SMILES: CCOC(=O)C[Si](C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)