missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Palmitoleate, TRC

Product Code. 30011396
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011396 1 g 1g
30006846 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011396 Supplier Toronto Research Chemicals Supplier No. TRCE9258551G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56219-10-4
Molecular Formula C18 H34 O2
Molecular Weight (g/mol) 282.46
Synonym (9Z)-9-Hexadecenoic Acid Ethyl Ester, (Z)-9-Hexadecenoic Acid Ethyl Ester, Palmitoleic Acid Ethyl Ester, Ethyl (Z)-9-Hexadecenoate, Ethyl cis-9-Hexadecenoate, cis-9-Hexadecenoic Acid Ethyl Ester
IUPAC Name ethyl (Z)-hexadec-9-enoate
SMILES CCCCCC\C=C/CCCCCCCC(=O)OCC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H34O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-4-2/h9-10H,3-8,11-17H2,1-2H3/b10-9-
Formula Weight 282.2559
Chemical Name or Material Ethyl Palmitoleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.