accessibility menu, dialog, popup

UserName

CAS RN 56219-10-4

H3COOCH3(Z)

CAS RN 56219-10-4

IUPAC Name: ethyl (9Z)-hexadec-9-enoate
Synonyms: ethyl (9Z)-hexadec-9-enoate
Molecular Weight (g/mol): 282.47
Molecular Formula: C18H34O2
InChi Key: JELGPLUONQGOHF-KTKRTIGZSA-N
SMILES: CCCCCC\C=C/CCCCCCCC(=O)OCC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results