missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,5-bis(trifluoromethyl)phenylacetate, 98%, Thermo Scientific™

Product Code. 11405540
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11405540 25 g 25g
11485530 1 g 1g
11495530 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405540 Supplier Thermo Scientific Alfa Aesar Supplier No. H25820.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144632-97-3
Molecular Formula C12H10F6O2
Molecular Weight (g/mol) 300.2
MDL Number MFCD08689894
InChI Key ICTVPOAERFKXSS-UHFFFAOYSA-N
Synonym ethyl 3,5-bis trifluoromethyl phenyl acetate, ethyl 3,5-bis trifluoromethyl phenylacetate, 3,5-bis trifluoromethyl phenylacetic acid ethyl ester, ethyl 2-3,5-bis trifluoromethyl phenyl acetate, ethyl 2-3,5-bis trifluoromethyl phenyl ethanoate, 3,5-di trifluoromethyl phenylacetic acid ethyl ester, 2-3,5-bis trifluoromethyl phenyl acetic acid ethyl ester
PubChem CID 42552891
IUPAC Name ethyl 2-[3,5-bis(trifluoromethyl)phenyl]acetate
SMILES CCOC(=O)CC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Refractive Index 1.417
Quantity 25 g
Formula Weight 300.2
Percent Purity 98%
Chemical Name or Material Ethyl 3,5-bis(trifluoromethyl)phenylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.