accessibility menu, dialog, popup

UserName

H3COOFFFFFF

CAS RN 144632-97-3

IUPAC Name: ethyl 2-[3,5-bis(trifluoromethyl)phenyl]acetate
Synonyms: ethyl 2-[3,5-bis(trifluoromethyl)phenyl]acetate
Molecular Weight (g/mol): 300.2
Molecular Formula: C12H10F6O2
InChi Key: ICTVPOAERFKXSS-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results