Learn More
Ethyl 2-isocyanato-4-(methylthio)butyrate, 97%, Thermo Scientific™
Product Code. 11397856
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397856 2 g 2g
11307866 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11397856 Supplier Thermo Scientific Alfa Aesar Supplier No. L13059.04
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 64505-12-0
Molecular Formula C8H13NO3S
Molecular Weight (g/mol) 203.256
MDL Number MFCD00078433
InChI Key IWXRXGRHXXLVHJ-UHFFFAOYSA-N
Synonym ethyl 2-isocyanato-4-methylthio butyrate, ethyl 2-isocyanato-4-methylthio butanoate, ethyl alpha-isocyanato gamma-thiomethyl butyrate, ethyl 2-isocyanato-4-methylthiobutyrate, ethyl 2-isocyanato-4-methylsulfanyl butanoate, butanoic acid,2-isocyanato-4-methylthio-, ethyl ester
PubChem CID 351396
IUPAC Name ethyl 2-isocyanato-4-methylsulfanylbutanoate
SMILES CCOC(=O)C(CCSC)N=C=O
Specifikationer
Boiling Point 155°C (24 mmHg)
Refractive Index 1.478
Quantity 2 g
UN Number UN2206
Beilstein 1710840
Sensitivity Moisture sensitive
Formula Weight 203.26
Percent Purity 97%
Chemical Name or Material Ethyl 2-isocyanato-4-(methylthio)butyrate
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.