accessibility menu, dialog, popup

UserName

CAS RN 64505-12-0

IUPAC Name: ethyl 2-isocyanato-4-(methylsulfanyl)butanoate
Synonyms: ethyl 2-isocyanato-4-(methylsulfanyl)butanoate
Molecular Weight (g/mol): 203.26
Molecular Formula: C8H13NO3S
InChi Key: IWXRXGRHXXLVHJ-UHFFFAOYNA-N
SMILES: CCOC(=O)C(CCSC)N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results