missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-(2-chlorophenyl)thiophene-3-carboxylate, 96%, Thermo Scientific™

Product Code. 15478345
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15478345 5 g 5g
15468345 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478345 Supplier Thermo Scientific Alfa Aesar Supplier No. H33171.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 325724-66-1
Molecular Formula C13H12ClNO2S
Molecular Weight (g/mol) 281.75
MDL Number MFCD01924157
InChI Key DDLXYLVCBHLQAW-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-2-chlorophenyl thiophene-3-carboxylate, 2-amino-4-2-chloro-phenyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylicacid, 2-amino-4-2-chlorophenyl-, ethyl ester, ethyl 2-amino-4-2-chlorophenyl-3-thiophenecarboxylate, ethyl 2-amino-4-2-chlorophenyl thiophene-2 carboxylate, 2-amino-4-2-chlorophenyl-thiophene-3-carboxylic acid ethyl ester, 2-amino-4-2-chlorophenyl thiophene-3-carboxylic acid ethyl ester
PubChem CID 1990440
IUPAC Name ethyl 2-amino-4-(2-chlorophenyl)thiophene-3-carboxylate
SMILES CCOC(=O)C1=C(N)SC=C1C1=CC=CC=C1Cl

Specifications

Melting Point 71°C to 73°C
Quantity 5 g
Formula Weight 281.76
Percent Purity 96%
Chemical Name or Material Ethyl 2-amino-4-(2-chlorophenyl)thiophene-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.