accessibility menu, dialog, popup

UserName

H3COOSClH2N

CAS RN 325724-66-1

IUPAC Name: ethyl 2-amino-4-(2-chlorophenyl)thiophene-3-carboxylate
Synonyms: ethyl 2-amino-4-(2-chlorophenyl)thiophene-3-carboxylate
Molecular Weight (g/mol): 281.75
Molecular Formula: C13H12ClNO2S
InChi Key: DDLXYLVCBHLQAW-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC=C1C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results