missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,2,3,3,3-pentafluoropropyl carbonate, 97%, Thermo Scientific™

Product Code. 15435157
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15435157

Brand: Thermo Scientific Alfa Aesar H32441.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 277332-96-4
Molecular Formula C6H7F5O3
Molecular Weight (g/mol) 222.111
MDL Number MFCD08688581
InChI Key NBYIBDCXTMTKLX-UHFFFAOYSA-N
Synonym ethyl 2,2,3,3,3-pentafluoropropyl carbonate, 2,2,3,3,3-pentafluoropropyl ethoxyformate, ethyl-2,2,3,3,3-pentafluoropropyl carbonate, carbonic acid ethyl 2,2,3,3,3-pentafluoropropyl ester
PubChem CID 26597286
IUPAC Name ethyl 2,2,3,3,3-pentafluoropropyl carbonate
SMILES CCOC(=O)OCC(C(F)(F)F)(F)F

Specifications

Linear Formula CF3CF2CH2OCO2CH2CH3
Refractive Index 1.33
Quantity 5 g
Formula Weight 222.11
Percent Purity 97%
Chemical Name or Material Ethyl 2,2,3,3,3-pentafluoropropyl carbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.