CAS RN 277332-96-4
CAS RN 277332-96-4
IUPAC Name:
ethyl 2,2,3,3,3-pentafluoropropyl carbonate
Synonyms:
ethyl 2,2,3,3,3-pentafluoropropyl carbonate
Molecular Weight (g/mol):
222.11
Molecular Formula:
C6H7F5O3
InChi Key:
NBYIBDCXTMTKLX-UHFFFAOYSA-N
SMILES:
CCOC(=O)OCC(F)(F)C(F)(F)F
1
–
1
of
1
results
1
Ethyl 2,2,3,3,3-pentafluoropropyl carbonate, 97%, Thermo Scientific™
CAS: 277332-96-4 Molecular Formula: C6H7F5O3 Molecular Weight (g/mol): 222.111 MDL Number: MFCD08688581 InChI Key: NBYIBDCXTMTKLX-UHFFFAOYSA-N Synonym: ethyl 2,2,3,3,3-pentafluoropropyl carbonate,2,2,3,3,3-pentafluoropropyl ethoxyformate,ethyl-2,2,3,3,3-pentafluoropropyl carbonate,carbonic acid ethyl 2,2,3,3,3-pentafluoropropyl ester PubChem CID: 26597286 IUPAC Name: ethyl 2,2,3,3,3-pentafluoropropyl carbonate SMILES: CCOC(=O)OCC(C(F)(F)F)(F)F