missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 1H-indazole-4-carboxylate, 95%

Product Code. 15474657
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15474657 250 mg 250mg
15464657 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474657 Supplier Alfa Aesar™ Supplier No. H31995.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 885279-45-8
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.202
MDL Number MFCD07371611
InChI Key ZYDRWPMGDIWCRN-UHFFFAOYSA-N
Synonym 1h-indazole-4-carboxylic acid ethyl ester, ethyl1h-indazole-4-carboxylate, ksc493g7d
PubChem CID 45790507
IUPAC Name ethyl 1H-indazole-4-carboxylate
SMILES CCOC(=O)C1=C2C=NNC2=CC=C1

Specifications

Melting Point 82°C to 85°C
Quantity 250 mg
Formula Weight 190.2
Percent Purity 95%
Chemical Name or Material Ethyl 1H-indazole-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.