accessibility menu, dialog, popup

UserName

CH3OONHN

CAS RN 885279-45-8

IUPAC Name: ethyl 1H-indazole-4-carboxylate
Synonyms: ethyl 1H-indazole-4-carboxylate
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: ZYDRWPMGDIWCRN-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C2C=NNC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results